Mal-PEG1-NHS ester ≥97%

Supplier: Ambeed
Warning

Synonyms: 2,5-Dioxopyrrolidin-1-yl 3-(2-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)ethoxy)propanoate

A741387-1G A741387-250MG A741387-5G
77573-374EA 477.92 USD
77573-374 77598-872 77622-654
Mal-PEG1-NHS ester ≥97%
Mal-PEG1-NHS ester

Precaución: for Research Use Only

Formula: C₁₃H₁₄N₂O₇
MW: 310.259 g/mol
MDL Number: MFCD26127783
CAS Number: 1807518-72-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR