5-Bromo-1-(phenylsulfonyl)-1H-pyrrolo[2,3-b]pyridine ≥98%

Supplier: Ambeed
Warning

A740408-5G A740408-1G A740408-250MG
77669-990EA 80 USD
77669-990 77873-170 77873-171
5-Bromo-1-(phenylsulfonyl)-1H-pyrrolo[2,3-b]pyridine ≥98%
5-Bromo-1-(phenylsulfonyl)-1H-pyrrolo[2,3-b]pyridine

Precaución: for Research Use Only

Formula: C₁₃H₉BrN₂O₂S
MDL Number: MFCD08741546
CAS Number: 1001070-33-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR