b-Amino-4-chlorobenzeneethanol ≥98%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-2-(4-chlorophenyl)ethanol

A740313-250MG A740313-100MG A740313-1G
77614-282EA 41.2 USD
77614-282 77684-666 77684-667
b-Amino-4-chlorobenzeneethanol ≥98%
b-Amino-4-chlorobenzeneethanol

Precaución: For research use only.

Formula: C₈H₁₀ClNO
MW: 171.624 g/mol
MDL Number: MFCD09759235
CAS Number: 179811-64-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR