4'-Carbamoyl-[1,1'-biphenyl]-4-yl methyl(3-(pyridin-4-yl)benzyl)carbamate ≥99%

Supplier: Ambeed
Warning

Synonyms: WWL70

A739931-1MG A739931-10MG A739931-5MG
77614-280EA 40.17 USD
77614-280 77686-748 77686-749
4'-Carbamoyl-[1,1'-biphenyl]-4-yl methyl(3-(pyridin-4-yl)benzyl)carbamate ≥99%
4'-Carbamoyl-[1,1'-biphenyl]-4-yl methyl(3-(pyridin-4-yl)benzyl)carbamate

Precaución: for Research Use Only

Formula: C₂₇H₂₃N₃O₃
MW: 437.49 g/mol
MDL Number: MFCD10567112
CAS Number: 947669-91-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR