6-Boc-2-oxo-6-azaspiro[3.5]nonane ≥98%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 2-oxo-6-azaspiro[3.5]nonane-6-carboxylate

A739644-100MG A739644-1G A739644-250MG
77882-622EA 54 USD
77882-622 77882-623 77882-624
6-Boc-2-oxo-6-azaspiro[3.5]nonane ≥98%
6-Boc-2-oxo-6-azaspiro[3.5]nonane

Precaución: for Research Use Only

Formula: C₁₃H₂₁NO₃
MW: 239.311 g/mol
MDL Number: MFCD21603789
CAS Number: 1359704-84-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR