(2S,4R)-1-((S)-2-(2-(2-Aminoethoxy)acetamido)-3,3-dimethylbutanoyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide ≥97%

Supplier: Ambeed
Warning

Synonyms: (S,R,S)-AHPC-PEG1-NH2

A739289-1G A739289-250MG A739289-50MG
77622-648EA 914.64 USD
77622-648 77622-650 77622-652
(2S,4R)-1-((S)-2-(2-(2-Aminoethoxy)acetamido)-3,3-dimethylbutanoyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide ≥97%
(2S,4R)-1-((S)-2-(2-(2-Aminoethoxy)acetamido)-3,3-dimethylbutanoyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide

Precaución: for Research Use Only

Formula: C₂₆H₃₇N₅O₅S
MW: 531.67 g/mol
MDL Number: MFCD32173010
CAS Number: 2241641-57-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR