7-Fluoro-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-6-boronic acid pinacol ester ≥95%

Supplier: Ambeed
Warning

Synonyms: 2H-1,4-Benzoxazin-3(4H)-one, 7-fluoro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-, 7-Fluoro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2H-benzo[b][1,4]oxazin-3(4H)-one, 7-Fluoro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2H-1,4-benzoxazin-3(4H)-one

A738898-1G A738898-250MG
77783-829EA 216 USD
77783-829 77783-830
7-Fluoro-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-6-boronic acid pinacol ester ≥95%
7-Fluoro-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-6-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₁₇BFNO₄
MDL Number: MFCD23098822
CAS Number: 2018286-71-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR