4-Acetamido-3-fluorobenzeneboronic acid, pinacol ester ≥98%

Supplier: Ambeed
Warning

A738607-5G A738607-1G
77891-464EA 630 USD
77891-464 77891-463
4-Acetamido-3-fluorobenzeneboronic acid, pinacol ester ≥98%
4-Acetamido-3-fluorobenzeneboronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₁₉BFNO₃
MDL Number: MFCD09878424
CAS Number: 269410-27-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR