3-Amino-5-fluoro-1H-indazole ≥98%

Supplier: Ambeed
Warning

A738149-1G A738149-250MG
77878-584EA 145 USD
77878-584 77878-585
3-Amino-5-fluoro-1H-indazole ≥98%
3-Amino-5-fluoro-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₆FN₃
MDL Number: MFCD03426699
CAS Number: 61272-72-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR