(3S,3aR,4S,4aR,7aR,8R,9aR)-3,4a,8-Trimethyl-2,5-dioxo-2,3,3a,4,4a,5,7a,8,9,9a-decahydroazuleno[6,5-b]furan-4-yl methacrylate ≥97%

Supplier: Ambeed
Warning

Synonyms: Arnicolide D

A737788-5MG A737788-1MG
77749-497EA 281 USD
77749-497 77749-496
(3S,3aR,4S,4aR,7aR,8R,9aR)-3,4a,8-Trimethyl-2,5-dioxo-2,3,3a,4,4a,5,7a,8,9,9a-decahydroazuleno[6,5-b]furan-4-yl methacrylate ≥97%
(3S,3aR,4S,4aR,7aR,8R,9aR)-3,4a,8-Trimethyl-2,5-dioxo-2,3,3a,4,4a,5,7a,8,9,9a-decahydroazuleno[6,5-b]furan-4-yl methacrylate

Precaución: for Research Use Only

Formula: C₁₉H₂₄O₅
MW: 332.39 g/mol
MDL Number: MFCD30730103
CAS Number: 34532-68-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR