Decyl-β-D-glucopyranoside 98+%, Ambeed.Inc

Supplier: Ambeed
Warning

A737536-1G A737536-5G A737536-250MG
77614-272EA 63.37 USD
77614-272 77684-664 77684-663
Decyl-β-D-glucopyranoside 98+%, Ambeed.Inc
Decyl-β-D-glucopyranoside

Precaución: For research use only.

Formula: C₁₆H₃₂O₆
MDL Number: MFCD00063297
CAS Number: 58846-77-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR