5,5',6,6',7,7',8,8'-Octahydro-[1,1'-binaphthalene]-2,2'-diol ≥98%

Supplier: Ambeed
Warning

A737174-1G A737174-25G A737174-5G
77614-266EA 30.9 USD
77614-266 77614-268 77614-270
5,5',6,6',7,7',8,8'-Octahydro-[1,1'-binaphthalene]-2,2'-diol ≥98%
5,5',6,6',7,7',8,8'-Octahydro-[1,1'-binaphthalene]-2,2'-diol

Precaución: for Research Use Only

Formula: C₂₀H₂₂O₂
MW: 294.39 g/mol
MDL Number: MFCD02093485
CAS Number: 39648-74-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR