(Z)-5-(3-(Methylamino)propylidene)-10,11-dihydro-5H-dibenzo[a,d][7]annulen-10-ol ≥99%

Supplier: Ambeed
Warning

Synonyms: (Z)-10-Hydroxynortriptyline

A736895-1MG A736895-5MG
77747-455EA 87 USD
77747-455 77747-456
(Z)-5-(3-(Methylamino)propylidene)-10,11-dihydro-5H-dibenzo[a,d][7]annulen-10-ol ≥99%
(Z)-5-(3-(Methylamino)propylidene)-10,11-dihydro-5H-dibenzo[a,d][7]annulen-10-ol

Precaución: for Research Use Only

Formula: C₁₉H₂₁NO
MW: 279.38 g/mol
CAS Number: 47132-19-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR