(3-Dibenzothiophen-4-ylphenyl)boronic acid ≥95%
Supplier: Ambeed
|
Warning
|
Synonyms:
(3-(Dibenzo[b,d]thiophen-4-yl)phenyl)boronic acid
A736749-1G
A736749-250MG
A736749-5G
77891-457EA
21
USD
77891-457
77891-458
77891-459
(3-Dibenzothiophen-4-ylphenyl)boronic acid ≥95%
(3-Dibenzothiophen-4-ylphenyl)boronic acid
Precaución: for Research Use Only
Formula:
C₁₈H₁₃BO₂S |
MDL Number:
MFCD22200801 CAS Number: 1307859-67-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...