4-Cyano-3-fluorophenyl-4-n-pentylbenzoate ≥98%

Supplier: Ambeed
Warning

Synonyms: 4-Pentylbenzoic acid 4-Cyano-3-fluorophenyl ester, 4-Amylbenzoic acid 4-Cyano-3-fluorophenyl ester, 4-Cyano-3-fluorophenyl 4-Pentylbenzoate, 4-Cyano-3-fluorophenyl 4-Amylbenzoate

A736086-1G A736086-5G
77669-982EA 24 USD
77669-982 77669-984
4-Cyano-3-fluorophenyl-4-n-pentylbenzoate ≥98%
4-Cyano-3-fluorophenyl-4-n-pentylbenzoate

Precaución: for Research Use Only

Formula: C₁₉H₁₈FNO₂
MW: 311.356 g/mol
MDL Number: MFCD11053363
CAS Number: 86786-89-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR