N-(9-((6aR,8R,9R,9aS)-9-Hydroxy-2,2,4,4-tetraisopropyltetrahydro-6H-furo[3,2-f][1,3,5,2,4]trioxadisilocin-8-yl)-6-oxo-6,9-dihydro-1H-purin-2-yl)isobutyramide ≥97%

Supplier: Ambeed
Warning

A736057-5G A736057-1G
77618-024EA 224.54 USD
77618-024 77749-494
N-(9-((6aR,8R,9R,9aS)-9-Hydroxy-2,2,4,4-tetraisopropyltetrahydro-6H-furo[3,2-f][1,3,5,2,4]trioxadisilocin-8-yl)-6-oxo-6,9-dihydro-1H-purin-2-yl)isobutyramide ≥97%
N-(9-((6aR,8R,9R,9aS)-9-Hydroxy-2,2,4,4-tetraisopropyltetrahydro-6H-furo[3,2-f][1,3,5,2,4]trioxadisilocin-8-yl)-6-oxo-6,9-dihydro-1H-purin-2-yl)isobutyramide

Precaución: for Research Use Only

Formula: C₂₆H₄₅N₅O₇Si₂
MW: 595.83 g/mol
MDL Number: MFCD09910466
CAS Number: 87865-78-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR