Ethyl 2,2-Difluoro-2-(trimethylsilyl)acetate ≥97%, Ambeed.Inc

Supplier: Ambeed
Danger

Synonyms: Ethyl 2,2-difluoro-2-(trimethylsilyl)acetate, 2,2-Difluoro-2-(trimethylsilyl)acetic acid Ethyl ester

A735037-1G A735037-25G A735037-5G
77684-658EA 43 USD
77684-658 77891-450 77891-451
Ethyl 2,2-Difluoro-2-(trimethylsilyl)acetate ≥97%, Ambeed.Inc
Ethyl 2,2-Difluoro-2-(trimethylsilyl)acetate

Precaución: For research use only.

Formula: C₇H₁₄F₂O₂Si
MDL Number: MFCD04973092
CAS Number: 205865-67-4
UN: 2924
ADR: 3, 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR