tert-Butyl 6-(hydroxymethyl)-2-azaspiro[3.3]heptane-2-carboxylate ≥95%

Supplier: Ambeed
Warning

A734760-100MG A734760-250MG
77686-739EA 98 USD
77686-739 77686-740
tert-Butyl 6-(hydroxymethyl)-2-azaspiro[3.3]heptane-2-carboxylate ≥95%
tert-Butyl 6-(hydroxymethyl)-2-azaspiro[3.3]heptane-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₂₁NO₃
MW: 227.3 g/mol
MDL Number: MFCD22419350
CAS Number: 1363381-93-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR