2-Nitro-5-(phenylsulfonyl)phenyl 4-chlorobenzoate ≥98%

Supplier: Ambeed
Danger

Synonyms: BTB06584

A734422-10MG A734422-1MG A734422-5MG
77617-996EA 47.38 USD
77617-996 77617-998 77618-000
2-Nitro-5-(phenylsulfonyl)phenyl 4-chlorobenzoate ≥98%
2-Nitro-5-(phenylsulfonyl)phenyl 4-chlorobenzoate

Precaución: for Research Use Only

Formula: C₁₉H₁₂ClNO₆S
MW: 417.82 g/mol
MDL Number: MFCD00103060
CAS Number: 219793-45-0
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR