Zinc meso-tetraphenylporphine ≥95%

Supplier: Ambeed
Warning

A734219-100MG A734219-250MG
77873-158EA 23 USD
77873-158 77873-159
Zinc meso-tetraphenylporphine ≥95%
Zinc meso-tetraphenylporphine

Precaución: for Research Use Only

Formula: C₄₄H₂₈N₄Zn
MDL Number: MFCD00012155
CAS Number: 14074-80-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR