Benzyl 4-(2-hydroxyethyl)piperidine-1-carboxylate ≥98%

Supplier: Ambeed
Warning

Synonyms: Z-4-(2-hydroxy-ethyl)piperidine

A734210-1G A734210-250MG A734210-25G A734210-5G
77684-654EA 86 USD
77684-654 77684-655 77684-656 77684-657
Benzyl 4-(2-hydroxyethyl)piperidine-1-carboxylate ≥98%
Benzyl 4-(2-hydroxyethyl)piperidine-1-carboxylate

Precaución: For research use only.

Formula: C₁₅H₂₁NO₃
MW: 263.33 g/mol
MDL Number: MFCD04114976
CAS Number: 115909-91-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR