5-Chloro-2-(methylsulfanyl)phenylboronic acid ≥98%

Supplier: Ambeed
Warning

Synonyms: 5-Chloro-2-(methythio)phenylboronic acid

A734099-100MG A734099-1G A734099-250MG
77901-481EA 47 USD
77901-481 77901-958 77901-833
5-Chloro-2-(methylsulfanyl)phenylboronic acid ≥98%
5-Chloro-2-(methylsulfanyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₈BClO₂S
MW: 202.47 g/mol
MDL Number: MFCD22543736
CAS Number: 1451392-55-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR