(R)-2-Bromo-3-phenylpropionic acid ≥97%

Supplier: Ambeed
Warning

A734080-5G A734080-1G
77674-008EA 446 USD
77674-008 77882-614
(R)-2-Bromo-3-phenylpropionic acid ≥97%
(R)-2-Bromo-3-phenylpropionic acid

Precaución: for Research Use Only

Formula: C₉H₉BrO₂
MDL Number: MFCD00210120
CAS Number: 42990-55-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR