N-(3-((2-(4-Chlorophenyl)-5-methyloxazol-4-yl)methoxy)benzyl)-N-(methoxycarbonyl)glycine 98+%

Supplier: Ambeed
Warning

Synonyms: BMS-687453

A734068-10MG A734068-1MG A734068-5MG
77617-988EA 49.44 USD
77617-988 77617-990 77617-992
N-(3-((2-(4-Chlorophenyl)-5-methyloxazol-4-yl)methoxy)benzyl)-N-(methoxycarbonyl)glycine 98+%
N-(3-((2-(4-Chlorophenyl)-5-methyloxazol-4-yl)methoxy)benzyl)-N-(methoxycarbonyl)glycine

Precaución: for Research Use Only

Formula: C₂₂H₂₁ClN₂O₆
MW: 444.87 g/mol
MDL Number: MFCD18251482
CAS Number: 1000998-59-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR