2-(2-(tert-Butoxycarbonyl)-2-azaspiro[3.3]heptan-6-yl)acetic acid ≥98%

Supplier: Ambeed
Warning

Synonyms: 2-{2-[(tert-butoxy)carbonyl]-2-azaspiro[3.3]heptan-6-yl}acetic acid

A733195-1G A733195-100MG A733195-250MG
77573-356EA 369.77 USD
77573-356 77614-254 77614-256
2-(2-(tert-Butoxycarbonyl)-2-azaspiro[3.3]heptan-6-yl)acetic acid ≥98%
2-(2-(tert-Butoxycarbonyl)-2-azaspiro[3.3]heptan-6-yl)acetic acid

Precaución: for Research Use Only

Formula: C₁₃H₂₁NO₄
MW: 255.31 g/mol
MDL Number: MFCD17016195
CAS Number: 1251002-39-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR