(3aS,4S,6aR)-4-(5-Hydroxypentyl)tetrahydro-1H-thieno[3,4-d]imidazol-2(3H)-one ≥96%

Supplier: Ambeed
Warning

Synonyms: D-Biotinol

A733034-100MG A733034-250MG A733034-1G A733034-5G
77598-864EA 57.4 USD
77598-864 77598-866 77622-628 77622-630
(3aS,4S,6aR)-4-(5-Hydroxypentyl)tetrahydro-1H-thieno[3,4-d]imidazol-2(3H)-one ≥96%
(3aS,4S,6aR)-4-(5-Hydroxypentyl)tetrahydro-1H-thieno[3,4-d]imidazol-2(3H)-one

Precaución: for Research Use Only

Formula: C₁₀H₁₈N₂O₂S
MW: 230.3271 g/mol
MDL Number: MFCD30532025
CAS Number: 53906-36-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR