Diethyl 3-fluorophthalate ≥95%, Ambeed.Inc

Supplier: Ambeed
Warning

A733009-1G A733009-250MG
77684-653EA 31 USD
77684-653 77891-449
Diethyl 3-fluorophthalate ≥95%, Ambeed.Inc
Diethyl 3-fluorophthalate

Precaución: For research use only.

Formula: C₁₂H₁₃FO₄
MDL Number: MFCD16037329
CAS Number: 65610-10-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR