(2S,4R)-N-((S)-2-(tert-Butyl)-17-((S)-4-(4-chlorophenyl)-2,3,9-trimethyl-6H-thieno[3,2-f][1,2,4]triazolo[4,3-a][1,4]diazepin-6-yl)-4,16-dioxo-6,9,12-trioxa-3,15-diazaheptadecan-1-oyl)-4-hydroxy-1-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide ≥98%

Supplier: Ambeed
Warning

Synonyms: MZ1

A732871-250MG A732871-100MG A732871-1MG A732871-5MG
77598-860EA 566.83 USD
77598-860 77622-620 77622-622 77622-624
(2S,4R)-N-((S)-2-(tert-Butyl)-17-((S)-4-(4-chlorophenyl)-2,3,9-trimethyl-6H-thieno[3,2-f][1,2,4]triazolo[4,3-a][1,4]diazepin-6-yl)-4,16-dioxo-6,9,12-trioxa-3,15-diazaheptadecan-1-oyl)-4-hydroxy-1-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide ≥98%
(2S,4R)-N-((S)-2-(tert-Butyl)-17-((S)-4-(4-chlorophenyl)-2,3,9-trimethyl-6H-thieno[3,2-f][1,2,4]triazolo[4,3-a][1,4]diazepin-6-yl)-4,16-dioxo-6,9,12-trioxa-3,15-diazaheptadecan-1-oyl)-4-hydroxy-1-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide

Precaución: for Research Use Only

Formula: C₄₉H₆₀ClN₉O₈S₂
MW: 1002.64 g/mol
MDL Number: MFCD30742945
CAS Number: 1797406-69-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR