(S)-(+)-2-Fluoro-α-methyl-4-biphenylacetic acid ≥98%

Supplier: Ambeed
Danger

A732143-1G A732143-250MG
77673-998EA 444.33 USD
77673-998 77674-000
(S)-(+)-2-Fluoro-α-methyl-4-biphenylacetic acid ≥98%
(S)-(+)-2-Fluoro-α-methyl-4-biphenylacetic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₃FO₂
MDL Number: MFCD00866152
CAS Number: 51543-39-6
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR