Dicyclohexylamine 2-cyanoacrylate ≥95%

Supplier: Ambeed
Warning

A731939-1G A731939-100MG
77573-350EA 154.5 USD
77573-350 77684-651
Dicyclohexylamine 2-cyanoacrylate ≥95%
Dicyclohexylamine 2-cyanoacrylate

Precaución: For research use only.

Formula: C₁₆H₂₆N₂O₂
MW: 278.38 g/mol
MDL Number: MFCD27920710
CAS Number: 263703-32-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR