4-[3-(Trifluoromethyl)phenyl]-4-piperidinol ≥95%

Supplier: Ambeed
Warning

A731611-100MG A731611-1G A731611-250MG A731611-5G
77627-480EA 27.81 USD
77627-480 77878-579 77878-580 77878-581
4-[3-(Trifluoromethyl)phenyl]-4-piperidinol ≥95%
4-[3-(Trifluoromethyl)phenyl]-4-piperidinol

Precaución: for Research Use Only

Formula: C₁₂H₁₄F₃NO
MDL Number: MFCD00044832
CAS Number: 2249-28-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR