2-Bromo-6-fluoro-3-hydroxyphenylboronic acid ≥95%

Supplier: Ambeed
Warning

Synonyms: (2-Bromo-6-fluoro-3-hydroxyphenyl)boronic acid

A731507-1G A731507-250MG A731507-5G
77884-649EA 253 USD
77884-649 77884-650 77884-651
2-Bromo-6-fluoro-3-hydroxyphenylboronic acid ≥95%
2-Bromo-6-fluoro-3-hydroxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₅BBrFO₃
MDL Number: MFCD16295246
CAS Number: 1451392-83-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR