H-Arg(Pbf)-OMe·HCl ≥98%
Supplier: Ambeed
|
Warning
|
Synonyms:
H-Arg(Pbf)-OMe·HCl, (S)-Methyl 2-amino-5-(3-((2,2,4,6,7-pentamethyl-2,3-dihydrobenzofuran-5-yl)sulphonyl)guanidino)pentanoate hydrochloride
A730949-100MG
A730949-250MG
77684-649EA
27
USD
77684-649
77684-650
H-Arg(Pbf)-OMe·HCl ≥98%
H-Arg(Pbf)-OMe·HCl
Precaución: For research use only.
Formula:
C₂₀H₃₃ClN₄O₅S MW: 477.017 g/mol |
MDL Number:
MFCD01318755 CAS Number: 257288-19-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...