3-Decenoic acid ≥90%

Supplier: Ambeed
Warning

A730813-100MG A730813-1G A730813-250MG
77617-978EA 17.51 USD
77617-978 77686-730 77686-731
3-Decenoic acid ≥90%
3-Decenoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₈O₂
MDL Number: MFCD00051464
CAS Number: 15469-77-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR