Cellotriose 98+%

Supplier: Ambeed
Warning

Synonyms: (2R,3R,4R,5R)-4-(((2S,3R,4R,5S,6R)-3,4-Dihydroxy-6-(hydroxymethyl)-5-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-2-yl)oxy)-2,3,5,6-tetrahydroxyhexanal, Glc beta(1-4)Glc beta(1-4)Glc, D-(+)-Cellotriose

A730235-50MG A730235-5MG
77627-474EA 722.03 USD
77627-474 77627-476
Cellotriose 98+%
Cellotriose

Precaución: for Research Use Only

Formula: C₁₈H₃₂O₁₆
MW: 504.44 g/mol
MDL Number: MFCD00069842
CAS Number: 33404-34-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR