2-Methoxynaphthalene-5-boronic acid ≥95%

Supplier: Ambeed
Warning

A730046-1G A730046-250MG A730046-100MG
77783-804EA 716 USD
77783-804 77783-805 77783-803
2-Methoxynaphthalene-5-boronic acid ≥95%
2-Methoxynaphthalene-5-boronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₁BO₃
MW: 202.01 g/mol
MDL Number: MFCD18412866
CAS Number: 1630264-42-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR