3-Methyl-5-(trifluoromethyl)benzoic acid ≥99%

Supplier: Ambeed
Warning

A729520-5G A729520-1G
77617-974EA 83.43 USD
77617-974 77749-482
3-Methyl-5-(trifluoromethyl)benzoic acid ≥99%
3-Methyl-5-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₉H₇F₃O₂
MDL Number: MFCD01631590
CAS Number: 117186-02-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR