1-Chloro-4-fluoro-3-iodobenzene ≥97%

Supplier: Ambeed
Warning

A729304-100G A729304-10G A729304-25G A729304-5G
77878-573EA 187 USD
77878-573 77878-574 77878-575 77878-576
1-Chloro-4-fluoro-3-iodobenzene ≥97%
1-Chloro-4-fluoro-3-iodobenzene

Precaución: for Research Use Only

Formula: C₆H₃ClFI
MDL Number: MFCD00672963
CAS Number: 116272-42-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR