2-(Bicyclo[2.2.1]heptan-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98+%

Supplier: Ambeed
Warning

A729059-1G A729059-250MG A729059-5G
77886-669EA 228 USD
77886-669 77886-670 77886-671
2-(Bicyclo[2.2.1]heptan-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98+%
2-(Bicyclo[2.2.1]heptan-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₃H₂₃BO₂
MDL Number: MFCD29919445
CAS Number: 174090-32-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR