3-Acetylpyridine adenine dinucleotide 90+%

Supplier: Ambeed
Warning

A728814-250MG A728814-100MG
77749-481EA 91 USD
77749-481 77749-480
3-Acetylpyridine adenine dinucleotide 90+%
3-Acetylpyridine adenine dinucleotide

Precaución: for Research Use Only

Formula: C₂₂H₂₈N₆O₁₄P₂
MDL Number: MFCD00078882
CAS Number: 86-08-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR