3-Hydroxy-5-(trifluoromethyl)phenylboronic acid ≥98%

Supplier: Ambeed
Warning

A728485-10G A728485-1G A728485-250MG A728485-5G
77886-665EA 734 USD
77886-665 77886-666 77886-667 77886-668
3-Hydroxy-5-(trifluoromethyl)phenylboronic acid ≥98%
3-Hydroxy-5-(trifluoromethyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₆BF₃O₃
MDL Number: MFCD09878337
CAS Number: 957062-66-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR