5-Chloro-1-benzothiophene-2-carbaldehyde ≥98%

Supplier: Ambeed
Warning

A728165-100MG A728165-250MG
77891-438EA 55 USD
77891-438 77891-439
5-Chloro-1-benzothiophene-2-carbaldehyde ≥98%
5-Chloro-1-benzothiophene-2-carbaldehyde

Precaución: for Research Use Only

Formula: C₉H₅ClOS
MDL Number: MFCD16622780
CAS Number: 28540-51-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR