(1-Ethyl-1H-benzo[d]imidazol-5-yl)boronic acid ≥98%

Supplier: Ambeed
Warning

A726782-250MG A726782-100MG A726782-1G
77622-616EA 85.49 USD
77622-616 77783-799 77783-800
(1-Ethyl-1H-benzo[d]imidazol-5-yl)boronic acid ≥98%
(1-Ethyl-1H-benzo[d]imidazol-5-yl)boronic acid

Precaución: for Research Use Only

Formula: C₉H₁₁BN₂O₂
MW: 190.01 g/mol
MDL Number: MFCD27935540
CAS Number: 1661043-48-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR