2-(2-Chloro-6-fluorophenyl)ethanol ≥95%

Supplier: Ambeed
Warning

A726326-1G A726326-5G
77669-958EA 21.49 USD
77669-958 77873-142
2-(2-Chloro-6-fluorophenyl)ethanol ≥95%
2-(2-Chloro-6-fluorophenyl)ethanol

Precaución: for Research Use Only

Formula: C₈H₈ClFO
MDL Number: MFCD00800609
CAS Number: 214262-86-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR