Clindamycin phosphate ≥758 µg/mg

Supplier: Ambeed
Warning

A726105-1G A726105-25G A726105-5G A726105-100G A726105-100MG
77602-874EA 6.15 USD
77602-874 77673-990 77673-992 77882-602 77882-603
Clindamycin phosphate ≥758 µg/mg
Clindamycin phosphate

Precaución: for Research Use Only

Formula: C₁₈H₃₄ClN₂O₈PS
MDL Number: MFCD07793328
CAS Number: 24729-96-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR