2,5-Dimethylpyridin-3-amine ≥95%

Supplier: Ambeed
Danger

A725801-100MG A725801-250MG
77886-658EA 33 USD
77886-658 77886-659
2,5-Dimethylpyridin-3-amine ≥95%
2,5-Dimethylpyridin-3-amine

Precaución: for Research Use Only

Formula: C₇H₁₀N₂
MDL Number: MFCD13175353
CAS Number: 89943-02-2
UN: 3259
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR