Tetrasodium-N,N-bis(carboxymethyl)-L-glutamate 40-48% in water
Supplier: Ambeed
|
Danger
|
Synonyms:
Sodium (S)-2-(bis(carboxylatomethyl)amino)pentanedioate, N,N-Bis(carboxymethyl)-L-glutamic Acid Tetrasodium Salt, Tetrasodium glutamate diacetate
A725659-5G
A725659-25G
77602-870EA
14.35
USD
77602-870
77669-956
Tetrasodium-N,N-bis(carboxymethyl)-L-glutamate 40-48% in water
Tetrasodium-N,N-bis(carboxymethyl)-L-glutamate
Precaución: for Research Use Only
Formula:
C₉H₁₃NO₈ |
MDL Number:
MFCD01862262 CAS Number: 51981-21-6 UN: 1760 ADR: 8,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...