3-Bromo-5-fluoro-2-nitropyridine ≥95%

Supplier: Ambeed
Warning

A725465-5G A725465-1G
77673-988EA 1458 USD
77673-988 77884-638
3-Bromo-5-fluoro-2-nitropyridine ≥95%
3-Bromo-5-fluoro-2-nitropyridine

Precaución: for Research Use Only

Formula: C₅H₂BrFN₂O₂
MDL Number: MFCD18632742
CAS Number: 1258544-91-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR