Cresol red sodium salt indicator

Supplier: Ambeed
Warning

A725290-100G A725290-25G A725290-500G A725290-5G
77614-234EA 135.96 USD
77614-234 77614-236 77684-642 77684-643
Cresol red sodium salt indicator
Cresol red sodium salt

Precaución: For research use only.

Formula: C₂₁H₁₇NaO₅S
MW: 404.42 g/mol
MDL Number: MFCD00001618
CAS Number: 62625-29-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR