4-((2-(2-(2-(2-(4-Aminophenoxy)ethoxy)ethoxy)ethoxy)ethyl)amino)-2-(2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione ≥98%

Supplier: Ambeed
Warning

A725159-100MG A725159-50MG A725159-1G A725159-250MG
77783-789EA 401 USD
77783-789 77783-792 77783-790 77783-791
4-((2-(2-(2-(2-(4-Aminophenoxy)ethoxy)ethoxy)ethoxy)ethyl)amino)-2-(2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione ≥98%
4-((2-(2-(2-(2-(4-Aminophenoxy)ethoxy)ethoxy)ethoxy)ethyl)amino)-2-(2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione

Precaución: for Research Use Only

Formula: C₂₇H₃₂N₄O₈
MDL Number: MFCD30490723
CAS Number: 1818885-63-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR